missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CENPM His Protein

Product Code. 18356195
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
10µg
20µg
250µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356195 50 μg 50µg
18389603 250 μg 250µg
18321462 20 μg 20µg
18379023 10 μg 10µg
18377013 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18356195 Supplier Novus Biologicals™ Supplier No. NBP31511750UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 79019
Molecular Weight (g/mol) M.W. Theoretical: 22.1 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias bK250D10.2, C22orf18PANE1MGC861, CENP-Mchromosome 22 open reading frame 18, centromere protein M, ICEN39, Interphase centromere complex protein 39, Pane1, proliferation associated nuclear element 1, Proliferation-associated nuclear element protein 1
Gene Symbol CENPM
Research Category DNA replication Transcription Translation and Splicing
Purity or Quality Grade >85%, by SDS-PAGE
Protein CENPM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.