missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CEACAM-6/CD66c Protein

Product Code. 18389467
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389467 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389467 Supplier R&D Systems™ Supplier No. 3934CM050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by the ability of the immobilized protein to support the adhesion of calcium ionophore treated human neutrophils. When 2 x 105 cells/well are added to CEACAM-6 coated plates (10 μg/mL, 100 μL/well), 35-60% of the cells will adhere after 20 minutes at 37° C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q53XP7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4680
Molecular Weight (g/mol) M.W. (Observed): 57-75 kDa, reducing conditions; M.W. (theoretical): 32.0 kDa
Name CEACAM-6/CD66c
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human CEACAM-6/CD66c protein Lys35-Gly320, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CEACAM6
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CEACAM6/CD66c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.