missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK4 Protein
A DNA sequence encoding the Homo sapiens (Human) CDK4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10035-ba-20ug
Additional Details : Weight : 0.01000kg
Especificaciones
CDK4 Protein | |
Research Use Only | |
CDK4 | |
Recombinant Protein | |
Baculovirus | |
MATSRYEPVA EIGVGAYGTV YKARDPHSGH FVALKSVRVP NGGGGGGGLP ISTVREVALL RRLEAFEHPN VVRLMDVCAT SRTDREIKRC FVESLSSVET LKPTSWAKSL T | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |