missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDH2 Protein

A cDNA sequence encoding the CDH2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11369-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937048

  • £5730.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CDH2 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADPDWVIPPI NLPENSRGPF PQELVRIRSD RDKNLSLRYS VTGPGADQPP TGIFIINPIS GQLSVTKPLD REQIARFHLR AHAVDINGNQ VENPIDIVIN VIDMNDNRPE FLHQVWNGTV PEGSKPGTYV MTVTAIDADD PNALNGMLRY RIVSQAPSTP SPNMFTINNE TGDIITVAAG LDREKVQQYT LIIQATDMEG NPTYGLSNTA TAVITVTDVN DNPPEFTAMT FYGEVPENRV DIIVANLTVT DKDQPHTPAW NAVYRISGGD PTGRFAIQTD PNSNDGLVTV VKPIDFETNR MFVLTVAAEN QVPLAKGIQH PPQSTATVSV TVIDVNENPY FAPNPKIIRQ EEGLHAGTML TTFTAQDPDR YMQQNIRYTK LSDPANWLKI DPVNGQITTI AVLDRESPNV KNNIYNATFL ASDNGIPPMS GTGTLQIYLL DINDNAPQVL PQEAETCETP DPNSINITAL DYDIDPNAGP FAFDLPLSPV TIKRNWTITR LNGDFAQLNL KIKFLEAGIY EVPIIITDSG NPPKSNISIL RVKVCQCDSN GDCTDVDRIV GAGLGTGAHH HHHH
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CDH2 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers