missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CDCP1 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
When Recombinant Human CDCP1 Fc Chimera (Catalog # 10402-CU) is immobilized at 1 μg/mL (100 μL/well), Recombinant Human P-Cadherin Fc Chimera (Catalog # 861-PC ) binds with an ED50 of 0.3-2.7 μg/mL.
Specifications
Specifications
| Accession Number | Q9H5V8.3 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 64866 |
| Molecular Weight (g/mol) | Observed: 110-130 kDa, under reducing conditions; Theroretical: 98 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CDCP1 protein Human CDCP1 (Phe30-Leu666) Accession # Q9H5V8.3(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction