missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDCA8 Protein

A cDNA sequence encoding the CDCA8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11366-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957038

  • £99.15 / 2µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CDCA8 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRKLASFLKD FDREVEIRIK QIESDRQNLL KEVDNLYNIE ILRLPKALRE MNWLDYFALG GNKQALEEAA TADLDITEIN KLTAEAIQTP LKSAKTRKVI QVDEMIVEEE EEEENERKNL QTARVKRCPP SKKRTQSIQG KGKGKRSSRA NTVTPAVGRLEVSMVKPTPG LTPRFDSRVF KTPGLRTPAA GERIYNISGN GSPLADSKEI FLTVPVGGGE SLRLLASDLQ RHSIAQLDPE ALGNIKKLSN RLAQICSSIR THK
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CDCA8 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 1mM DTT, 1mM EDTA and 0.2M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers