missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CDC25B (aa 377-566) Protein

Product Code. 18375317
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375317 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375317 Supplier R&D Systems™ Supplier No. 1649CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by hydrolysis of 250 μM 3-O-methyl fluorescein phosphate (OMFP). The specific activity is >0.5 μmol/min/mg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004349
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, EDTA, DTT and Glycerol.
Gene ID (Entrez) 994
Molecular Weight (g/mol) M.W. (Observed): 23-25 kDa, reducing conditions; M.W. (theoretical): 23 kDa
Name CDC25B aa 377-566
Quantity 50 μg
Source E. coli-derived human CDC25B protein Glu377-Gln566, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Based on mass spectrometric analysis, a carboxyl-terminal truncated peptide with a molecular mass of approximately 22.1 kDa may also be present in the recombinant protein preparation.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol CDC25B
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CDC25B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.