missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDA Protein
A cDNA sequence encoding the CDA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10546-1mg
Additional Details : Weight : 0.01000kg
Specifications
978 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity :> 3.5unit/mg. One unit of activity was defined as the amount required to deaminate 1.0 umole of cytidine per min at pH 7.5 at 250C. | |
Human | |
His | |
The CDA solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 8.0), 1mM DTT, 2mM EDTA, 100mM NaCl and 40% Glycerol. |
CDA Protein | |
Research Use Only | |
CDA | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAQKRPACTL KPECVQQLLV CSQEAKQSAY CPYSHFPVGA ALLTQEGRIF KGCNIENACY PLGICAERTA IQKAVSEGYK DFRAIAIASD MQDDFISPCG ACRQVMREFG TNWPVYMTKP DGTYIVMTVQ ELLPSSFGPE DLQKTQ | |
Greater than 90.0% as determined by SDS-PAGE. |