missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD8B Protein
A cDNA sequence encoding the CD8B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11353-HIS-20ug
Additional Details : Weight : 0.01000kg
Specifications
926 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLQQTPAY IKVQTNKMVM LSCEAKISLS NMRIYWLRQR QAPSSDSHHE FLALWDSAKG TIHGEEVEQE KIAVFRDASR FILNLTSVKP EDSGIYFCMI VGSPELTFGK GTQLSVVDFL PTTAQPTKKS TLKKRVCRLP RPETQKGPLC SP | |
Greater than 80% as determined by SDS-PAGE. |
CD8B Protein | |
Research Use Only | |
CD8B | |
Human | |
His | |
The CD8B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |