missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD74 Protein

A cDNA sequence encoding the CD74 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11346-HIS-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936958

  • £99.15 / 5µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

972
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQQQGRLD KLTVTSQNLQ LENLRMKLPK PPKPVSKMRM ATPLLMQALP MGALPQGPMQ NATKYGNMTE DHVMHLLQNA DPLKVYPPLK GSFPENLRHL KNTMETIDWK VFESWMHHWL LFEMSRHSLE QKPTDAPPKE SLELEDPSSG LGVTKQDLGP VPM
Greater than 90.0% as determined by SDS-PAGE.
CD74 Protein
Research Use Only
CD74
Human
His
CD74 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers