missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD73 / NT5E Protein
A DNA sequence enoding full length human CD73 plus a c-terminal 6x histidine tag was expressed in human HEK293 cells.
Brand: enQuireBio™ QP2192-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191742.1 | |
CD73 / NT5E Protein | |
Research Use Only | |
NT5E | |
Measured by its ability to hydrolyze the 5'-phosphate group from the substrate adenosine 5'-monophosphate (AMP). The specific activity is > 20,000 pmoles/min/ug. | |
Human | |
His | |
0.22 um filtered solution of TBS plus 4mM CaCl2 and 20% Glycerol at pH 7.5 |
4907 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
plus a c-terminal 6x histidine tag was expressed in human HEK293 cells. | |
Recombinant Protein | |
Human | |
WELTILHTND VHSRLEQTSE DSSKCVNASR CMGGVARLFT KVQQIRRAEP NVLLLDAGDQ YQGTIWFTVY KGAEVAHFMN ALRYDAMALG NHEFDNGVEG LIEPLLKEAK FPILSANIKA KGPLASQISG LYLPYKVLPV GDEVVGIVGY TSKETPFLSN PGTNLVFEDE ITALQPEVDK LKTLNVNKII ALGHSGFEMD KLIAQKVRGV DVVVGGHSNT FLYTGNPPSK EVPAGKYPFI VTSDDGRKVP VVQAYAFGKY LGYLKIEFDE RGNVISSHGN PILLNSSIPE DPSIKADINK WRIKLDNYST QELGKTIVYL DGSSQSCRFR ECNMGNLICD AMINNNLRHA DETFWNHVSM CILNGGGIRS PIDERNNGTI TWENLAAVLP FGGTFDLVQL KGSTLKKAFE HSVHRYGQST GEFLQVGGIH VVYDLSRKPG DRVVKLDVLC TKCRVPSYDP LKMDEVYKVI LPNFLANGGD GFQMIKDELL RHDSGDQDIN VVSTYISKMK VIYPAVEGRI KVD | |
> 95 % as determined by SDS-PAGE |