missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD73 / NT5E Protein

A DNA sequence enoding full length human CD73 plus a c-terminal 6x histidine tag was expressed in human HEK293 cells.

Brand:  enQuireBio™ QP2192-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965539

  • £201.00 / 10µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001191742.1
CD73 / NT5E Protein
Research Use Only
NT5E
Measured by its ability to hydrolyze the 5'-phosphate group from the substrate adenosine 5'-monophosphate (AMP). The specific activity is > 20,000 pmoles/min/ug.
Human
His
0.22 um filtered solution of TBS plus 4mM CaCl2 and 20% Glycerol at pH 7.5
4907
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
plus a c-terminal 6x histidine tag was expressed in human HEK293 cells.
Recombinant Protein
Human
WELTILHTND VHSRLEQTSE DSSKCVNASR CMGGVARLFT KVQQIRRAEP NVLLLDAGDQ YQGTIWFTVY KGAEVAHFMN ALRYDAMALG NHEFDNGVEG LIEPLLKEAK FPILSANIKA KGPLASQISG LYLPYKVLPV GDEVVGIVGY TSKETPFLSN PGTNLVFEDE ITALQPEVDK LKTLNVNKII ALGHSGFEMD KLIAQKVRGV DVVVGGHSNT FLYTGNPPSK EVPAGKYPFI VTSDDGRKVP VVQAYAFGKY LGYLKIEFDE RGNVISSHGN PILLNSSIPE DPSIKADINK WRIKLDNYST QELGKTIVYL DGSSQSCRFR ECNMGNLICD AMINNNLRHA DETFWNHVSM CILNGGGIRS PIDERNNGTI TWENLAAVLP FGGTFDLVQL KGSTLKKAFE HSVHRYGQST GEFLQVGGIH VVYDLSRKPG DRVVKLDVLC TKCRVPSYDP LKMDEVYKVI LPNFLANGGD GFQMIKDELL RHDSGDQDIN VVSTYISKMK VIYPAVEGRI KVD
> 95 % as determined by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers