missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD7 Protein

A DNA sequence encoding the Homo sapiens (Human) CD7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5794-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947809

  • £380.58 / 100µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006128.1
CD7 Protein
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
924
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD7
Recombinant Protein
E. coli
AQEVQQSPHC TTVPVGASVN ITCSTSGGLR GIYLRQLGPQ PQDIIYYEDG VVPTTDRRFR GRIDFSGSQD NLTITMHRLQ LSDTGTYTCQ AITEVNVYGS GTLVLVTEEQ SQGWHRCSDA PPRASALPAP PTGSALPDPQ TASALPDPPA ASALP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers