missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD58 Protein

A cDNA sequence encoding the CD58 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11340-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936928

  • £155.00 / 20µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

965
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFSQQIYG VVYGNVTFHV PSNVPLKEVL WKKQKDKVAE LENSEFRAFS SFKNRVYLDT VSGSLTIYNL TSSDEDEYEM ESPNITDTMK FFLYVLESLP SPTLTCALTN GSIEVQCMIP EHYNSHRGLI MYSWDCPMEQ CKRNSTSIYF KMENDLPQKI QCTLSNPLFN TTSSIILTTC IPSSGHSRHR
Greater than 90.0% as determined by SDS-PAGE.
CD58 Protein
Research Use Only
CD58
Human
His
CD58 protein solution (1 mg/ml) containing 20mM Phosphate buffer and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers