missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD5 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human CD5 Fc Chimera (Catalog # 11389-CD) binds to Human CD5 Antibody (Catalog # MAB16361 ) with a ED50 of 0.800-8.00 ng/mL.
Specifications
Specifications
| Accession Number | P06127.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 921 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 72-83 kDa, under reducing conditions., MolecularWeight-theroretical: 65 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD5 protein Human CD5 (Arg25-Asn371) (N-terminus) Accession # P06127.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD5, CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction