missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD44 / HCAM Protein
A DNA sequence encoding the Homo sapiens (Human) CD44 / HCAM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5788-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000601.3 | |
CD44 / HCAM Protein | |
Research Use Only | |
CD44 | |
Recombinant Protein | |
E. coli | |
QIDLNITCRF AGVFHVEKNG RYSISRTEAA DLCKAFNSTL PTMAQMEKAL SIGFETCRYG FIEGHVVIPR IHPNSICAAN NTGVYILTSN TSQYDTYCFN ASAPPEEDCT SVTDLPNAFD GPITITIVNR DGTRYVQKGE YRTNPEDIYP SNPTDDDVSS GSSSERSSTS GGYIFYTFST VHPIPDEDSP WITDSTDRIP ATSTSSNTIS AGWEPNEENE DERDRHLSFS GSGIDDDEDF ISSTISTTPR AFDHTKQNQD WTQWNPSHSN PEVLLQTTTR MTDVDRNGTT AYEGNWNPEA HPPLIHHEHH EEEETPHSTS TIQATPSSTT EETATQKEQW FGNRWHEGYR QTPKEDSHST TGTAAASAHT SHPMQGRTTP SPEDSSWTDF FNPISHPMGR GHQAGRRMDM DSSHSITLQP TANPNTGLVE DLDRTGPLSM TTQQSNSQSF STSHEGLEED KDHPTTSTLT SSNRNDVTGG RRDPNHSEGS TTLLEGYTSH YPHTKESRTF IPVTSAKTGS FGVTAVTVGD SNSNVNRSLS GDQDTFHPSG GSHTTHGSES DGHSHGSQEG GANTTSGPIR TPQIPE | |
Greater than 90% as determined by SDS-PAGE. |
960 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |