missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD34 Protein

Product Code. 15976878
Click to view available options
Quantity:
1 mg
10 μg
2 μg
20 μg
5 μg
Unit Size:
10µg
1mg
20µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15976878

Brand: enQuireBio™ QP11328HIS5ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CD34 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 947
Name CD34 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol CD34
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MLVRRGARAG PRMPRGWTAL CLLSLLPSGF MSLDNNGTAT PELPTQGTFS NVSTNVSYQE TTTPSTLGST SLHPVSQHGN EATTNITETT VKFTSTSVIT SVYGNTNSSV QSQTSVISTV FTTPANVSTP ETTLKPSLSP GNVSDLSTTS TSLATSPTKP YTSSSPILSD IKAEIKCSGI REVKLTQGIC LEQNKTSSCA EFKKDRGEGL ARVLCGEEQA DADAGAQVCS LLLAQSEVRP QCLLLVLANR TEISSKLQLM KKHQSDLKKL GILDFTEQDV ASHQSYSQKT LIALVTSGAL LAVLGITGYF LMNRRSWSPT GERLGEDPYY TENGGGQGYS SGPGTSPEAQ GKASVNRGAQ ENGTGQATSR NGHSARQHVV ADTEL
Buffer CD34 in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced).
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.