missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD300d Fc Chimera Protein

Product Code. 18797046
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18797046 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18797046 Supplier R&D Systems™ Supplier No. 10273CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect is 1 to 6 μg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UXZ3
For Use With (Application) Bioactivity
Gene ID (Entrez) 100131439
Molecular Weight (g/mol) 43 kDa
Name CD300 molecule-like family member d, CD300d, CD300LD, CLM-5, CMRF35-A4, immune receptor CD300d, MAIR-IV
Quantity 50 μg
Source

Chinese Hamster Ovary cell line, CHO-derived human CD300d protein

Human CD300d (Ala19-His165) Accession # Q6UXZ3IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.