missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD300A Protein

A cDNA sequence encoding the CD300A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11324-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936858

  • £155.00 / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

11314
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLSKCRTV AGPVGGSLSV QCPYEKEHRT LNKYWCRPPQ IFLCDKIVET KGSAGKRNGR VSIRDSPANL SFTVTLENLT EEDAGTYWCG VDTPWLRDFH DPVVEVEVSV FPAS
Greater than 90% as determined by SDS-PAGE.
CD300A Protein
Research Use Only
CD300A
Human
His
The CD300A solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers