missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD25/IL-2R alpha Fc Chimera Protein

Product Code. 18390378
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390378 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390378 Supplier R&D Systems™ Supplier No. 1020RL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CD25/IL-2 R alpha Fc Chimera Protein is derived from NS0. The Recombinant Human CD25/IL-2 R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3559
Molecular Weight (g/mol) 48.3 kDa (monomer)
Name CD25/IL-2R alpha
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human CD25/IL-2 R alpha protein, (N-terminus) Human CD25/IL-2 R alpha (Glu22-Cys213) Accession &Num; Q5W007 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.