missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD1E Protein
A cDNA sequence encoding the CD1E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11311-1mg
Additional Details : Weight : 0.01000kg
Specifications
913 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEEQLSFR MLQTSSFANH SWAHSEGSGW LGDLQTHGWD TVLGTIRFLK PWSHGNFSKQ ELKNLQSLFQ LYFHSFIQIV QASAGQFQLE YPFEIQILAG CRMNAPQIFL NMAYQGSDFL SFQGISWEPS PGAGIRAQNI CKVLNRYLDI KEILQSLLGH TCPRFLAGLM EAGESELKRK VKPEAWLSCG PSPGPGRLQL VCHVSGFYPK PVWVMWMRGE QEQRGTQRGD VLPNADETWY LRATLDVAAG EAAGLSCRVK HSSLGGHDLI IHWGGYS | |
Greater than 85.0% as determined by SDS-PAGE. |
CD1E Protein | |
Research Use Only | |
CD1E | |
Human | |
His | |
CD1E protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |