missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD1D Protein
A cDNA sequence encoding the CD1D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11310-50ug
Additional Details : Weight : 0.01000kg
Specifications
CD1D Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
EVPQRLFPLR CLQISSFANS SWTRTDGLAW LGELQTHSWS NDSDTVRSLK PWSQGTFSDQ QWETLQHIFR VYRSSFTRDV KEFAKMLRLS YPLELQVSAG CEVHPGNASN NFFHVAFQGK DILSFQGTSW EPTQEAPLWV NLAIQVLNQD KWTRETVQWL LNGTCPQFVS GLLESGKSELKKQVKPKAWL SRGPSPGPGR LLLVCHVSGF YPKPVWVKWM RGEQEQQGTQ PGDILPNADE TWYLRATLDV VAGEAAGLSC RVKHSSLEGQ DIVLYWGGSY TSLEHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD1D protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |