missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD1A Protein

Product Code. 15986788
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 15986788

Brand: enQuireBio™ QP1130850ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CD1A was constructed and used to recombinantly synthesize the protein.

Specifications

Name CD1A Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence ADPDGLKEPL SFHVTWIASF YNHSWKQNLV SGWLSDLQTH TWDSNSSTIV FLCPWSRGNF SNEEWKELET LFRIRTIRSF EGIRRYAHEL QFEYPFEIQV TGGCELHSGK VSGSFLQLAY QGSDFVSFQN NSWLPYPVAG NMAKHFCKVL NQNQHENDIT HNLLSDTCPR FILGLLDAGK AHLQRQVKPE AWLSHGPSPG PGHLQLVCHV SGFYPKPVWV MWMRGEQEQQ GTQRGDILPS ADGTWYLRAT LEVAAGEAAD LSCRVKHSSL EGQDIVLYWE HHSSVHHHHH H
Buffer CD1A protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4), and 10% glycerol.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.