missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CD164L2 His Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantity:
0.1 mg
0.5 mg
Conditionnement:
0.1mg
0.5mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-141 of Human CD164L2 The Recombinant Human CD164L2 Protein is derived from E. coli. The Recombinant Human CD164L2 Protein has been validated for the following applications: SDS-Page.
Spécification
Spécification
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT |
| Gene ID (Entrez) | 388611 |
| Molecular Weight (g/mol) | TMW: 14.4kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CD164 Sialomucin-Like 2, CD164 Sialomucin-Like 2 Protein, EAPG6122, UNQ6122 |
| Common Name | CD164L2 |
| Conjugate | Unconjugated |
| Afficher plus |
For research use only and is not approved for use in humans or in clinical diagnosis
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu