missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD155 / PVR Protein

A DNA sequence encoding the Homo sapiens (Human) CD155 / PVR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6559-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074150

  • £384.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129240.1
CD155 / PVR Protein
Research Use Only
PVR
Recombinant Protein
E. coli
WPPPGTGDVV VQAPTQVPGF LGDSVTLPCY LQVPNMEVTH VSQLTWARHG ESGSMAVFHQ TQGPSYSESK RLEFVAARLG AELRNASLRM FGLRVEDEGN YTCLFVTFPQ GSRSVDIWLR VLAKPQNTAE VQKVQLTGEP VPMARCVSTG GRPPAQITWH SDLGGMPNTS QVPGFLSGTV TVTSLWILVP SSQVDGKNVT CKVEHESFEK PQLLTVNLTV YYPPEVSISG YDNNWYLGQN EATLTCDARS NPEPTGYNWS TTMGPLPPFA VAQGAQLLIR PVDKPINTTL ICNVTNALGA RQAELTVQVK EGPPSEHSGI SRN
Greater than 90% as determined by SDS-PAGE.
5817
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers