missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD155 / PVR Protein
A DNA sequence encoding the Homo sapiens (Human) CD155 / PVR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6559-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129240.1 | |
CD155 / PVR Protein | |
Research Use Only | |
PVR | |
Recombinant Protein | |
E. coli | |
WPPPGTGDVV VQAPTQVPGF LGDSVTLPCY LQVPNMEVTH VSQLTWARHG ESGSMAVFHQ TQGPSYSESK RLEFVAARLG AELRNASLRM FGLRVEDEGN YTCLFVTFPQ GSRSVDIWLR VLAKPQNTAE VQKVQLTGEP VPMARCVSTG GRPPAQITWH SDLGGMPNTS QVPGFLSGTV TVTSLWILVP SSQVDGKNVT CKVEHESFEK PQLLTVNLTV YYPPEVSISG YDNNWYLGQN EATLTCDARS NPEPTGYNWS TTMGPLPPFA VAQGAQLLIR PVDKPINTTL ICNVTNALGA RQAELTVQVK EGPPSEHSGI SRN | |
Greater than 90% as determined by SDS-PAGE. |
5817 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |