missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD14 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£278.61
Specifications
Gene ID (Entrez) | 929 |
---|---|
Molecular Weight (g/mol) | Observed M.W.: 43-58 kDa, reducing conditions |
Name | CD14 |
Quantity | 50μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human CD14 protein Thr20-Cys352 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18395627
|
R&D Systems™
383-CD-050/CF |
£278.61
50µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18167470
|
R&D Systems™
383-CD-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£278.61
50µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human CD14 Protein is derived from CHO. The Recombinant Human CD14 Protein has been validated for the following applications: Bioactivity.Specifications
929 | |
CD14 | |
Chinese Hamster Ovary cell line,CHO-derived human CD14 protein Thr20-Cys352 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Observed M.W.: 43-58 kDa, reducing conditions | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |