missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCNI Protein

Product Code. 15946758
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15946758 1 mg 1mg
15966758 5 μg 5µg
15956758 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15946758 Supplier enQuireBio™ Supplier No. QP112981mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CCNI was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 10983
Name CCNI Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol CCNI
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMKFPGPL ENQRLSFLLE KAITREAQMW KVNVRKMPSN QNVSPSQRDE VIQWLAKLKY QFNLYPETFA LASSLLDRFL ATVKAHPKYL SCIAISCFFL AAKTVEEDER IPVLKVLARD SFCGCSSSEI LRMERIILDK LNWDLHTATP LDFLHIFHAI AVSTRPQLLF SLPKLSPSQH LAVLTKQLLH CMACNQLLQF RGSMLALAMV SLEMEKLIPD WLSLTIELLQ KAQMDSSQLI HCRELVAHHL STLQSSLPLN SVYVYRPLKH TLVTCDKGVF RLHPSSVPGP DFSKDNSKPE VPVRGTAAFY HHLPAASGCK QTSTKRKVEE MEVDDFYDGI KRLYNEDNVS ENVGSVCGTD LSRQEGHASP CPPLQPVSVM
Buffer CCNI protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.