missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNI Protein
A cDNA sequence encoding the CCNI was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11298-20ug
Additional Details : Weight : 0.01000kg
Specifications
10983 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
CCNI | |
Human | |
His | |
CCNI protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
CCNI Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKFPGPL ENQRLSFLLE KAITREAQMW KVNVRKMPSN QNVSPSQRDE VIQWLAKLKY QFNLYPETFA LASSLLDRFL ATVKAHPKYL SCIAISCFFL AAKTVEEDER IPVLKVLARD SFCGCSSSEI LRMERIILDK LNWDLHTATP LDFLHIFHAI AVSTRPQLLF SLPKLSPSQH LAVLTKQLLH CMACNQLLQF RGSMLALAMV SLEMEKLIPD WLSLTIELLQ KAQMDSSQLI HCRELVAHHL STLQSSLPLN SVYVYRPLKH TLVTCDKGVF RLHPSSVPGP DFSKDNSKPE VPVRGTAAFY HHLPAASGCK QTSTKRKVEE MEVDDFYDGI KRLYNEDNVS ENVGSVCGTD LSRQEGHASP CPPLQPVSVM |