missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL8 / MCP-2 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL8 / MCP-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP4994-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_005614.2 | |
CCL8 / MCP-2 Protein | |
Research Use Only | |
CCL8 | |
Recombinant Protein | |
Yeast | |
QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP | |
Greater than 90% as determined by SDS-PAGE. |
6355 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |