missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL3L1 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/mL.
Specifications
Specifications
| Accession Number | P16619 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 6349 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CCL3L1/LD78 beta as determined by LAL method. |
| Gene Alias | C-C motif chemokine 3-like 1, CCL3L3, chemokine (C-C motif) ligand 3-like 1, D17S1718, G0/G1 switch regulatory protein 19-2, G0S19-2LD78, LD78BETA, LD78-beta(1-70), MGC104178, MGC12815, MGC182017, MIP1AP, PAT 464.2, SCYA3L, SCYA3L1464.2, small inducible cytokine A3-like 1, Small-inducible cytokine A3-like 1, Tonsillar lymphocyte LD78 beta protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?