missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL28 is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human lymphocytes is in a concentration range of 1.0-10.0 ng/mL.
Specifications
Specifications
| Accession Number | Q9NRJ3, Q9NRJ3, Q9NRJ3 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. |
| Gene ID (Entrez) | 56477 |
| Molecular Weight (g/mol) | 12.4 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CCL28 as determined by LAL method. |
| Gene Alias | chemokine (C-C motif) ligand 28, MEC/CCL28, member 28, MGC71902, Protein CCK1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?