missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human CCR3 transfected HEK293 cells is in a concentration range of 0.5-2.0 μg/mL.
Specifications
Specifications
| Accession Number | Q9Y258, Q9Y258, Q9Y258 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 10344 |
| Molecular Weight (g/mol) | 8.4 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CCL26/Eotaxin-3 as determined by LAL method. |
| Gene Alias | CC chemokine IMAC, C-C motif chemokine 26, CCL26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin-3/CCL26, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?