missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL22 / MDC Protein
A DNA sequence encoding the Homo sapiens (Human) CCL22 / MDC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8521-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002981.2 | |
CCL22 / MDC Protein | |
Research Use Only | |
CCL22 | |
Recombinant Protein | |
E. coli | |
GPYGANMEDS VCCRDYVRYR LPLRVVKHFY WTSDSCPRPG VVLLTFRDKE ICADPRVP | |
Greater than 90% as determined by SDS-PAGE. |
6367 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |