missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL21/6Ckine Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£373.64
Specifications
Gene ID (Entrez) | 6366 |
---|---|
Molecular Weight (g/mol) | 12 kDa |
Name | CCL21/6Ckine |
Quantity | 25μg |
Source | E. coli-derived human CCL21/6Ckine protein Ser24-Pro134 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18301708
|
R&D Systems™
366-6C-025 |
£373.64
25µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18167730
|
R&D Systems™
366-6C-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£373.64
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human CCL21/6Ckine Protein is derived from E. coli. The Recombinant Human CCL21/6Ckine Protein has been validated for the following applications: Bioactivity.Specifications
6366 | |
CCL21/6Ckine | |
E. coli-derived human CCL21/6Ckine protein Ser24-Pro134 | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
12 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |