missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL21/6Ckine (Mucin Stalk Chimera) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Human CCL21/6Ckine (Mucin Stalk Chimera) Protein is derived from NS0. The Recombinant Human CCL21/6Ckine (Mucin Stalk Chimera) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 6366 |
| Molecular Weight (g/mol) | 38.3 kDa |
| Name | CCL21/6Ckine Mucin-Like Stalk Chimera |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human CCL21/6Ckine protein, (N-terminus) Human 6Ckine (Ser24-Pro134) (Pro41 Ser) Accession &Num; NP_002980 GS Human Fractalkine mucin-like stalk (Phe103-Gln341) Accession &Num; P78423 KQN 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction