missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL17 / TARC / SCYA17 Protein

Product Code. 16018611
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
This item is not returnable. View return policy

Product Code. 16018611

Brand: enQuireBio™ QP97500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_002978.1
Gene ID (Entrez) 6361
Name CCL17 / TARC / SCYA17 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol CCL17
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.