missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL16 / HCC-4 / NCC4 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL16 / HCC-4 / NCC4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5771-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004581.1 | |
CCL16 / HCC-4 / NCC4 Protein | |
Research Use Only | |
CCL16 | |
Recombinant Protein | |
E. coli | |
QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ | |
Greater than 90% as determined by SDS-PAGE. |
6360 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |