missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MCP4 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration of 10-100 ng/mL.
Specifications
Specifications
| Accession Number | Q99616, Q99616, Q99616 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. |
| Gene ID (Entrez) | 6357 |
| Molecular Weight (g/mol) | 8.6 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CCL13/MCP-4 as determined by LAL method. |
| Gene Alias | C-C motif chemokine 13, CCL13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4/CCL13, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?