missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL11/Eotaxin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£255.93 - £569.10
Specifications
Gene ID (Entrez) | 6356 |
---|---|
Molecular Weight (g/mol) | 8.4 kDa |
Name | CCL11/Eotaxin |
Source | E. coli-derived human CCL11/Eotaxin protein Gly24-Pro97 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18339257
|
R&D Systems™
320-EO-020/CF |
£255.93
20µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18335327
|
R&D Systems™
320-EO-020 |
£255.93
20µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18116430
|
R&D Systems™
320-EO-100 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 100μg |
£569.10
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18117030
|
R&D Systems™
320-EO-100/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | 100μg |
£569.10
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Human CCL11/Eotaxin Protein is derived from E. coli. The Recombinant Human CCL11/Eotaxin Protein has been validated for the following applications: Bioactivity.Specifications
6356 | |
CCL11/Eotaxin | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
8.4 kDa | |
E. coli-derived human CCL11/Eotaxin protein Gly24-Pro97 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |