missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Cathepsin X/Z/P Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human Cathepsin X/Z/P Protein is derived from NS0. The Recombinant Human Cathepsin X/Z/P Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Acetate and NaCl. |
| Gene ID (Entrez) | 1522 |
| Molecular Weight (g/mol) | 33 kDa |
| Name | Cathepsin X/Z/P |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Cathepsin X/Z/P protein Ala21-Val303 (Gly23Val & Ser48Thr),with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Pro form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction