missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Cathepsin W His Protein

Product Code. 18126329 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
0.25 mg
Unit Size:
0.05mg
0.25mg
This item is not returnable. View return policy

Product Code. 18126329

Brand: Novus Biologicals™ NBP2515170.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-376 of Human Cathepsin W The Recombinant Human Cathepsin W Protein is derived from E. coli. The Recombinant Human Cathepsin W Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol
Gene ID (Entrez) 1521
Molecular Weight (g/mol) TMW: 42kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias cathepsin W, cathepsin W (lymphopain), EC 3.4.22.-, Lymphopain, LYPN
Common Name Cathepsin W
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.