missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Cathepsin F His Protein
Description
Specific activity is > 5 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1pmole of Z-Phe-Arg-AMC to Z-Phe-Arg and AMC per minute at pH 5.0 at 37.
Specifications
Specifications
| Concentration | 0.25 mg/ml |
| For Use With (Application) | Enzyme Activity, Functional Assay |
| Formulation | PBS (pH 7.4), 40% glycerol |
| Gene ID (Entrez) | 8722 |
| Molecular Weight (g/mol) | 52.5 kDa |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
| Gene Alias | cathepsin F, CATSFEC 3.4.22.41, EC 3.4.22 |
| Gene Symbol | CTSF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?