missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Caspase-9 Protein

Product Code. 18431511 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431511 25 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431511 Supplier Novus Biologicals™ Supplier No. NBP19960625Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE, Bioactivity
Formulation Semi Dry powder
Gene ID (Entrez) 842
Quantity 25 U
Storage Requirements Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbol CASP9
Reconstitution Reconstitute to 1 unit per l in PBS containing 15% glycerol.
Inhibitor Properties Each Definition: One unit of the recombinant caspase-9 is the enzyme activity that cleaves 1 nmol of the caspase substrate LEHD-pNA (pNA: pnitroanaline) per hour at 37°C in a reaction solution containing 50 mM Hepes pH 7.2 50 mM NaCl 0.1% Chaps 10 mM EDTA 5% Glycerol and 10 mM DTT. Specific activity: >400 units/mg
Research Category Apoptosis, Cancer, Caspases, Cell Biology
Purity or Quality Grade >90%, by SDS-PAGE
Protein Caspase-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.