missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonic anhydrase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonic anhydrase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5740-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001122301.1 | |
Carbonic anhydrase 1 Protein | |
Research Use Only | |
Ca1 | |
Recombinant Protein | |
E. coli | |
ASPDWGYDDK NGPEQWSKLY PIANGNNQSP VDIKTSETKH DTSLKPISVS YNPATAKEII NVGHSFHVNF EDNDNRSVLK GGPFSDSYRL FQFHFHWGST NEHGSEHTVD GVKYSAELHV AHWNSAKYSS LAEAASKADG LAVIGVLMKV GEANPKLQKV LDALQAIKTK GKRAPFTNFD PSTLLPSSLD FWTYPGSLTH PPLYESVTWI ICKESISVSS EQLAQFRSLL SNVEGDNAVP MQHNNRPTQP LKGRTVRASF | |
Greater than 90% as determined by SDS-PAGE. |
759 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |