missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CANX Protein

A cDNA sequence encoding the CANX was constructed and used to recombinantly synthesize the protein.

£98.39 - £1989.41

Specifications

Name CANX Protein
Product Type Recombinant Protein
Protein Tag Untagged
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15936678
View Documents Promotion Details
enQuireBio™
QP11271-10UG
10 μg
£98.39
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15956678
View Documents Promotion Details
enQuireBio™
QP11271-50UG
50 μg
£151.37
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15946678
View Documents Promotion Details
enQuireBio™
QP11271-1MG
1 mg
£1989.41
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CANX Protein
Untagged
Human
Greater than 90.0% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MHDGHDDDVI DIEDDLDDVI EEVEDSKPDT TAPPSSPKVT YKAPVPTGEV YFADSFDRGT LSGWILSKAK KDDTDDEIAK YDGKWEVEEM KESKLPGDKG LVLMSRAKHH AISAKLNKPF LFDTKPLIVQ YEVNFQNGIE CGGAYVKLLS KTPELNLDQF HDKTPYTIMF GPDKCGEDYK LHFIFRHKNP KTGIYEEKHA KRPDADLKTY FTDKKTHLYT LILNPDNSFE ILVDQSVVNS GNLLNDMTPP VNPSREIEDP EDRKPEDWDE RPKIPDPEAV KPDDWDEDAP AKIPDEEATK PEGWLDDEPE YVPDPDAEKP EDWDEDMDGE WEAPQIANPR CESAPGCGVW QRPVIDNPNY KGKWKPPMID NPSYQGIWKP RKIPNPDFFE DLEPFRMTPF SAIGLELWSM TSDIFFDNFI ICADRRIVDD WANDGWGLKK AADGAAEPGV VGQMIEAAEE RP
SDS
Product Certifications