missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CANT1 Protein
Description
Measured by its ability to hydrolyze the beta 5'-phosphate group from the substrate uridine-5'-diphosphate (UDP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996 ). The specific activity is >85,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q8WVQ1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 124583 |
| Molecular Weight (g/mol) | M.W. (Observed): 36-40 kDa, reducing conditions; M.W. (theoretical): 37 kDa |
| Name | Calcium Activated Nucleotidase 1/CANT1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human Calcium Activated Nucleotidase 1/CANT1 protein Gly80-Ile401, with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?