missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CaMKII beta GST (N-Term) Protein

Product Code. 18335298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335298 25 μg 25µg
18328447 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18335298 Supplier Novus Biologicals™ Supplier No. H00000816P0125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Tekniske data

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification, Kinase Assay
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 816
Molecular Weight (g/mol) M.W. (Theoretical): 81.07 kDa
Quantity 25 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II beta, calcium/calmodulin-dependent protein kinase II beta, calcium/calmodulin-dependent protein kinase type II beta chain, calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II beta subunit, CaM kinase II subunit beta, CAM2CAMK2, CAMKB, CaMK-II subunit beta, CaM-kinase II beta chain, EC 2.7.11, EC 2.7.11.17, MGC29528, proline rich calmodulin-dependent protein kinase
Gene Symbol CAMK2B
Research Category Protein Kinase, Wnt Signaling Pathway
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CaMKII beta
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.