missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALCB / CGPR / Calcitonin 2 Protein
A DNA sequence encoding the Homo sapiens (Human) CALCB / CGPR / Calcitonin 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5747-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000719.1 | |
CALCB / CGPR / Calcitonin 2 Protein | |
Research Use Only | |
CALCB | |
Recombinant Protein | |
E. coli | |
ACNTATCVTH RLAGLLSRSG GMVKSNFVPT NVGSKAF | |
Greater than 80% as determined by SDS-PAGE. |
797 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |