missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Cadherin-12 (CAD-12) Fc Chimera
Description
The Recombinant Human Cadherin-12 (CAD-12) Fc Chimera is derived from NS0. The Recombinant Human Cadherin-12 (CAD-12) Fc Chimera has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris-Citrate, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 1010 |
| Molecular Weight (g/mol) | 92.5 kDa (monomer) |
| Name | Pro Cadherin-12 Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Cadherin-12 protein, (N-terminus) Human Pro-Cadherin-12 (Gln24-Pro609) Accession &Num; P55289 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?